Learn More
Biomatik Corporation Interleukin 12A (Il12A) Monoclonal Antibody
Supplier:  Biomatik Corporation CAU29598
Encompass_Preferred
Interleukin 12A (Il12A) Monoclonal Antibody is a Human (Homo sapiens) Monoclonal Antibody to Interleukin 12A (IL12A). The Interleukin 12A (Il12A) Monoclonal Antibody has been validated for the following applications: WB. Host: Mouse. Species: Human (Homo sapiens). Immunogen: Arg23~Ser219. Applications: WB. Target Synonyms: Interleukin 12A (IL12A). Accession Number: P29459. Gene ID: 3592. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7582