Learn More
Cusabio Technology LLC interleukin 13 IL-13
Catalog No. 5000048910
Encompass_Preferred
This kit detects IL13 in guinea pig serum plasma and tissue homogenates using a quantitative sandwich ELISA method It provides a sensitive detection range of 7 5 pg/mL to 300 pg/mL with a minimum detectable concentration of 3 75 pg/mL ensuring accurate measurement of low-abundance samples The assay requires a sample volume of 50-100 L and can be completed within 1-5 hours offering flexibility for laboratory workflows Absorbance is measured at 450 nm and the kit is optimized for reliable performance in diverse biological matrices making it suitable for researchers analyzing IL13 levels in immunological or inflammatory studies
Catalog No. 50-000-48910 Supplier Cusabio Technology LLC Supplier No. CSBE12916GP
This kit detects IL13 in guinea pig serum plasma and tissue homogenates using a quantitative sandwich ELISA method It provides a sensitive detection range of 7 5 pg/mL to 300 pg/mL with a minimum detectable concentration of 3 75 pg/mL ensuring accurate measurement of low-abundance samples The assay requires a sample volume of 50-100 L and can be completed within 1-5 hours offering flexibility for laboratory workflows Absorbance is measured at 450 nm and the kit is optimized for reliable performance in diverse biological matrices making it suitable for researchers analyzing IL13 levels in immunological or inflammatory studies