Learn More
Cusabio Technology LLC Mouse Interleukin 13IL-13 ELISA Kit
Catalog No. NC3654191
Encompass_Preferred
The mouse IL13 (IL13) ELISA Kit quantitates mouse IL13 levels in multiple samples, including serum, plasma, and tissue homogenates. IL-13 is a Th2 cytokine involved in effective repair after acute lung injury and the pathogenesis of chronic diseases such as allergic asthma. IL-13 induces hypersensitivity of the airways, augments mucous production, activates fibroblasts, elevates serum Ig E levels, and increases the number of eosinophils. During schistosomiasis and other pathologies, IL-13 has been described to promote liver fibrosis. IL-13 binds to the type II receptor IL-4Rα and IL-13Rα1 for signaling via JAK1/2, leading to the activation of STAT6 and subsequent transcription of genes affecting cell proliferation and inflammation.
Catalog No. NC3654191 Supplier Cusabio Technology LLC Supplier No. CSBE04602M
Add to Cart
Add to Cart
The mouse IL13 (IL13) ELISA Kit quantitates mouse IL13 levels in multiple samples, including serum, plasma, and tissue homogenates. IL-13 is a Th2 cytokine involved in effective repair after acute lung injury and the pathogenesis of chronic diseases such as allergic asthma. IL-13 induces hypersensitivity of the airways, augments mucous production, activates fibroblasts, elevates serum Ig E levels, and increases the number of eosinophils. During schistosomiasis and other pathologies, IL-13 has been described to promote liver fibrosis. IL-13 binds to the type II receptor IL-4Rα and IL-13Rα1 for signaling via JAK1/2, leading to the activation of STAT6 and subsequent transcription of genes affecting cell proliferation and inflammation.