Learn More
Genscript Corporation IL-13 Variant, Human
Catalog No. 50276421
Encompass
The human IL-13 cDNA encodes a 132 amino acid protein containing a proposed 20 amino acid signal peptide. Human IL-13 shares approximately 30% amino acid sequence homology to human IL-4 and the two cytokines exhibit overlapping biological activities. Human IL-13 is produced by activated Th0, Th1-like Th2-like and CD8 T cells. Similarly to IL-4, IL-13 has multiple effects on the differentiation and functions of monocytes/macrophages. IL-13 can suppress the cytotoxic functions of monocytes/macrophages. It can also suppress the production of proinflammatory cytokines and upregulate the production of IL-1ra by monocytes/macrophages.
Catalog No. 50-276-421 Supplier Genscript Corporation Supplier No. Z0271110
The human IL-13 cDNA encodes a 132 amino acid protein containing a proposed 20 amino acid signal peptide. Human IL-13 shares approximately 30% amino acid sequence homology to human IL-4 and the two cytokines exhibit overlapping biological activities. Human IL-13 is produced by activated Th0, Th1-like Th2-like and CD8 T cells. Similarly to IL-4, IL-13 has multiple effects on the differentiation and functions of monocytes/macrophages. IL-13 can suppress the cytotoxic functions of monocytes/macrophages. It can also suppress the production of proinflammatory cytokines and upregulate the production of IL-1ra by monocytes/macrophages.