Learn More
Cayman Chemical InterleukIn-13humnrecomb 100ug
SDP
Catalog No. 502945324
Encompass_Preferred
Source: Active recombinant human IL-13 expressed in CHO cells • Amino acids: 21-132 • MW: 12.5 kDa
Catalog No. 50-294-5324 Supplier Cayman Chemical Supplier No. 40467100 UG
Source: Active recombinant human IL-13 expressed in CHO cells • Amino acids: 21-132 • MW: 12.5 kDa