Learn More
Creative Biomart Interleukin-16 (IL-16) Protein
Catalog No. 502084303
Encompass_Preferred
Interleukin-16 Human Recombinant produced in E.Coli is a single, non-glycosylated polypeptide chain containing 130 amino acids and having a molecular mass of 13.5 kDa. The IL-16 is purified by proprietary chromatographic techniques.
Catalog No. 50-208-4303 Supplier Creative Biomart Supplier No. THP0252
Interleukin-16 Human Recombinant produced in E.Coli is a single, non-glycosylated polypeptide chain containing 130 amino acids and having a molecular mass of 13.5 kDa. The IL-16 is purified by proprietary chromatographic techniques.