Learn More
Biomatik Corporation Interleukin 17 (Il17) Monoclonal Antibody
Supplier:  Biomatik Corporation CAU29157
Encompass_Preferred
Interleukin 17 (Il17) Monoclonal Antibody is a Rabbit (Oryctolagus cuniculus) Monoclonal Antibody to Interleukin 17 (IL17). The Interleukin 17 (Il17) Monoclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Mouse. Species: Rabbit (Oryctolagus cuniculus). Immunogen: Val21~Ala153. Applications: WB, IHC, ICC, IP. Target Synonyms: Interleukin 17 (IL17). Accession Number: G1SLF2. Gene ID: 100339322. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4828