Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Interleukin-17 receptor A protein | >95.0% | 2 UG
SDP

Catalog No. 5000291591
Encompass_Preferred

IL-17RA Protein, Rat (HEK293, His) is a recombinant form of rat interleukin-17 receptor A produced in HEK293 mammalian cells with a C-terminal His tag. The protein (M1-P320) is supplied as a purified preparation for use in biochemical characterization, binding studies, and cell-based assays.

  • Recombinant rat interleukin-17 receptor A for research applications.
  • Expressed in HEK293 cells for mammalian post-translational modifications.
  • C-terminal His tag facilitates purification and detection.
  • Protein length approximately 320 amino acids (M1-P320).
  • Purity greater than 95% by reducing SDS-PAGE.
  • Suitable for biochemical, binding, and cell-based assays.
  • Store at -20°C for stability.

Catalog No. 50-002-91591 Supplier Medchemexpress LLC Supplier No. HYP7482510UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

IL-17RA Protein, Rat (HEK293, His) is a recombinant form of rat interleukin-17 receptor A produced in HEK293 mammalian cells with a C-terminal His tag. The protein (M1-P320) is supplied as a purified preparation for use in biochemical characterization, binding studies, and cell-based assays.

  • Recombinant rat interleukin-17 receptor A for research applications.
  • Expressed in HEK293 cells for mammalian post-translational modifications.
  • C-terminal His tag facilitates purification and detection.
  • Protein length approximately 320 amino acids (M1-P320).
  • Purity greater than 95% by reducing SDS-PAGE.
  • Suitable for biochemical, binding, and cell-based assays.
  • Store at -20°C for stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.