Learn More
Cusabio Technology LLC interleukin-18 binding prorein
Supplier:  Cusabio Technology LLC CSBE17797M
Encompass_Preferred
This kit detects IL-18BP in mouse serum plasma and cell culture supernates using a quantitative sandwich ELISA method It has a detection range of 0 156 ng/mL to 10 ng/mL and a sensitivity of 0 039 ng/mL enabling accurate measurement of IL-18BP levels in diverse biological samples The assay requires 50-100 L of sample per test and can be completed within 1 to 5 hours Results are determined by measuring absorbance at 450 nm making it suitable for researchers requiring reliable quantification of IL-18BP in experimental or diagnostic applications
Catalog No. 50-000-49984