Learn More
Medchemexpress LLC Interleukin-18 receptor accessory protein (IL-18R beta) | 100 UG
SDP
Catalog No. 5000290208
Encompass_Preferred
Recombinant human interleukin-18 receptor accessory protein (IL-18R beta/IL-1R7) expressed in HEK293 cells and fused to an Fc tag. Supplied as a purified protein for in vitro research applications including receptor binding studies, signaling assays, and antibody validation.
  • Fc-tagged human protein expressed in HEK293 cells.
  • Suitable for receptor binding and signaling assays.
  • Purified recombinant protein for in vitro research use.
  • Accession O95256-1 and gene ID 8807 provide sequence reference.
  • Supplied as a 100 μg quantity for laboratory assays.
Catalog No. 50-002-90208 Supplier Medchemexpress LLC Supplier No. HYP76425100UG
Recombinant human interleukin-18 receptor accessory protein (IL-18R beta/IL-1R7) expressed in HEK293 cells and fused to an Fc tag. Supplied as a purified protein for in vitro research applications including receptor binding studies, signaling assays, and antibody validation.
  • Fc-tagged human protein expressed in HEK293 cells.
  • Suitable for receptor binding and signaling assays.
  • Purified recombinant protein for in vitro research use.
  • Accession O95256-1 and gene ID 8807 provide sequence reference.
  • Supplied as a 100 μg quantity for laboratory assays.