Learn More
Medchemexpress LLC Interleukin-18 receptor accessory protein (IL-18R beta, human, Fc fusion) | 50 UG
SDP
Catalog No. 5000290164
Encompass_Preferred
Recombinant human interleukin-18 receptor accessory protein (IL-18R beta) fused to an Fc tag, expressed in HEK293 cells and supplied as a lyophilized powder. Supplied quantity 50 μg; molecular weight 55-80 kDa; endotoxin <1 EU/μg. Formulated in PBS with 5% trehalose; recommended reconstitution ≥100 μg/mL. Store at -20°C; after reconstitution stable at 4°C for 1 week or frozen for long-term storage.
  • Fc-tagged construct facilitates purification and detection.
  • Human sequence expressed in mammalian cells for native glycosylation.
  • Lyophilized formulation improves shelf life and shipping stability.
  • Low endotoxin level suitable for sensitive assays.
  • Recommended reconstitution concentration preserves activity.
  • Suitable for biochemical and cell-based assays.
Catalog No. 50-002-90164 Supplier Medchemexpress LLC Supplier No. HYP7642550UG
Recombinant human interleukin-18 receptor accessory protein (IL-18R beta) fused to an Fc tag, expressed in HEK293 cells and supplied as a lyophilized powder. Supplied quantity 50 μg; molecular weight 55-80 kDa; endotoxin <1 EU/μg. Formulated in PBS with 5% trehalose; recommended reconstitution ≥100 μg/mL. Store at -20°C; after reconstitution stable at 4°C for 1 week or frozen for long-term storage.
  • Fc-tagged construct facilitates purification and detection.
  • Human sequence expressed in mammalian cells for native glycosylation.
  • Lyophilized formulation improves shelf life and shipping stability.
  • Low endotoxin level suitable for sensitive assays.
  • Recommended reconstitution concentration preserves activity.
  • Suitable for biochemical and cell-based assays.