Learn More
Medchemexpress LLC Interleukin-18 receptor accessory protein, recombinant human (Fc fusion) | >95.0% | 10 UG
SDP
Catalog No. 5000290560
Encompass_Preferred
Recombinant human interleukin-18 receptor accessory protein (IL-18R beta/IL-1R7) is supplied as an Fc-tagged fusion produced in HEK293 cells and provided lyophilized. The material is highly purified and formulated in PBS with 5% trehalose; reconstitution at or above 100 μg/mL is recommended. It is intended for receptor-binding, biochemical assays, and reagent development.
  • Fc fusion facilitates dimerization and detection
  • High purity (>95.0%) for biochemical and binding studies
  • Lyophilized formulation for extended shelf life
  • Reconstitute to ≥100 μg/mL in ddH2O as recommended
  • Store lyophilized at -20°C; aliquot and freeze for long-term storage
Catalog No. 50-002-90560 Supplier Medchemexpress LLC Supplier No. HYP7642510UG
Recombinant human interleukin-18 receptor accessory protein (IL-18R beta/IL-1R7) is supplied as an Fc-tagged fusion produced in HEK293 cells and provided lyophilized. The material is highly purified and formulated in PBS with 5% trehalose; reconstitution at or above 100 μg/mL is recommended. It is intended for receptor-binding, biochemical assays, and reagent development.
  • Fc fusion facilitates dimerization and detection
  • High purity (>95.0%) for biochemical and binding studies
  • Lyophilized formulation for extended shelf life
  • Reconstitute to ≥100 μg/mL in ddH2O as recommended
  • Store lyophilized at -20°C; aliquot and freeze for long-term storage