Learn More
Cusabio Technology LLC Interleukin 1B IL- 1B
Catalog No. 5000048361
Encompass_Preferred
The Fish Interleukin 1B (IL-1B) ELISA Kit is designed to quantitatively measure IL1B levels in serum plasma and tissue homogenates with high precision This kit detects IL1B within a broad dynamic range of 3 12 pg/mL to 200 pg/mL offering a sensitivity as low as 0 78 pg/mL Utilizing a sandwich ELISA methodology it ensures reliable and specific detection of the target protein The assay requires a short incubation period of 1-5 hours and uses a sample volume of 50-100 L per test Results are measured at an absorbance wavelength of 450 nm providing a straightforward workflow for researchers studying immune responses or inflammatory pathways in fish models Its robust performance makes it suitable for applications in both basic research and biomarker validation
Catalog No. 50-000-48361 Supplier Cusabio Technology LLC Supplier No. CSBE13259FH
The Fish Interleukin 1B (IL-1B) ELISA Kit is designed to quantitatively measure IL1B levels in serum plasma and tissue homogenates with high precision This kit detects IL1B within a broad dynamic range of 3 12 pg/mL to 200 pg/mL offering a sensitivity as low as 0 78 pg/mL Utilizing a sandwich ELISA methodology it ensures reliable and specific detection of the target protein The assay requires a short incubation period of 1-5 hours and uses a sample volume of 50-100 L per test Results are measured at an absorbance wavelength of 450 nm providing a straightforward workflow for researchers studying immune responses or inflammatory pathways in fish models Its robust performance makes it suitable for applications in both basic research and biomarker validation