Learn More
Sigma Aldrich Fine Chemicals Biosciences Interleukin 2 from mouse I
Catalog No. 502904372
Encompass
IL-2 (interleukin-2) codes for a pro-inflammatory cytokine. IL-2 is mostly expressed in T helper cells.
Catalog No. 50-290-4372 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. I052320UG
IL-2 (interleukin-2) codes for a pro-inflammatory cytokine. IL-2 is mostly expressed in T helper cells.