Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Interleukin 2, human

Catalog No. AAJ67298EXE
Change view
Click to view available options
Quantity:
10μg
1mg
50μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ67298EXE 10μg
AAJ67298IAM 50μg
AAJ67298MCR 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ67298EXE Supplier Thermo Scientific Chemicals Supplier No. J67298EXE

15.5 kd g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Molecular Weight (g/mol) 15.5 kd
Quantity 10μg
Packaging Plastic bottle
Form Lyophilized
How should I store reconstituted Interleukin 2, human (Cat. No. J67298.EXE)?

Reconstituted solutions of Interleukin 2, human are stable for up to one week at 2-8°C. For long-term storage of stock solutions, we recommend that you portion the stock into working aliquots and store at -80°C.

How should I reconstitute Interleukin 2, human (Cat. No. J67298.EXE)?

To reconstitue Interleukin 2, human, we recommend the following:

  • Briefly centrifuge the tube prior to opening it to bring the contents to the bottom
  • Use sterile water to reconstitute to a concentration of 0.1-1.0 mg/mL in a siliconized tube


Further dilutions should be made in appropriate buffered solutions.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.