Learn More
Sigma Aldrich Fine Chemicals Biosciences Interleukin-2 human hIL-2
Catalog No. 502905459
Encompass
Recombinant Interleukin-2 human (hIL-2) is produced in E. coli and purified by standard chromatographic techniques. SOLUTIONon in PBS and 1 mg/ml BSA filtered through a 0.2 mum pore size membrane.
Catalog No. 50-290-5459 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 10799068001
Recombinant Interleukin-2 human (hIL-2) is produced in E. coli and purified by standard chromatographic techniques. SOLUTIONon in PBS and 1 mg/ml BSA filtered through a 0.2 mum pore size membrane.