Learn More
Sigma Aldrich Fine Chemicals Biosciences Interleukin 2 human IL 2 r10UG
Catalog No. 502914067
Encompass
Interleukin-2 (IL-2) is an helical cytokine belonging to T-helper cell type 1. It has a molecular weight of 15kDa. The IL2 gene is mapped to human chromosome 4q27. IL-2 is originally called as T cell growth factor.
Catalog No. 50-291-4067 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. I26445X10UG
Interleukin-2 (IL-2) is an helical cytokine belonging to T-helper cell type 1. It has a molecular weight of 15kDa. The IL2 gene is mapped to human chromosome 4q27. IL-2 is originally called as T cell growth factor.