Learn More
Medchemexpress LLC Interleukin-2 receptor subunit beta (IL-2R beta/CD122) protein, canine (recombinant, HEK293 hFc) | >95.0% | 2 UG
SDP
Catalog No. 5000294095
Encompass_Preferred
Recombinant canine interleukin-2 receptor beta (IL-2Rβ/CD122) extracellular domain expressed in HEK293 cells and provided as a C-terminal human Fc fusion. Supplied as a lyophilized powder for receptor-ligand binding, immunoassays, and functional studies requiring the extracellular portion of canine IL-2Rβ.
  • Recombinant extracellular domain of canine IL-2Rβ for biologically relevant binding studies.
  • Expressed in HEK293 cells for mammalian glycosylation.
  • C-terminal human Fc fusion enhances stability and enables Fc-based detection.
  • Supplied lyophilized for long-term storage and convenient reconstitution.
  • High purity (>95.0%) suitable for biochemical and immunological assays.
  • Low endotoxin (<1 EU/μg) for sensitive cell-based experiments.
Catalog No. 50-002-94095 Supplier Medchemexpress LLC Supplier No. HYP767692UG
Recombinant canine interleukin-2 receptor beta (IL-2Rβ/CD122) extracellular domain expressed in HEK293 cells and provided as a C-terminal human Fc fusion. Supplied as a lyophilized powder for receptor-ligand binding, immunoassays, and functional studies requiring the extracellular portion of canine IL-2Rβ.
  • Recombinant extracellular domain of canine IL-2Rβ for biologically relevant binding studies.
  • Expressed in HEK293 cells for mammalian glycosylation.
  • C-terminal human Fc fusion enhances stability and enables Fc-based detection.
  • Supplied lyophilized for long-term storage and convenient reconstitution.
  • High purity (>95.0%) suitable for biochemical and immunological assays.
  • Low endotoxin (<1 EU/μg) for sensitive cell-based experiments.