Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Interleukin-20 receptor subunit beta (mouse) | 2 UG
SDP

Catalog No. 5000290210
Encompass_Preferred

Recombinant mouse interleukin-20 receptor subunit beta (IL-20RB) produced in HEK293 cells with a C-terminal His tag. IL-20RB forms functional heterodimers (for example with IL-20RA or IL-22RA1) and participates in STAT3 signaling; it serves as a receptor subunit for IL-19, IL-20, and IL-24. The protein is intended for biochemical characterization and cell-signaling assays.

  • Species: mouse
  • Expression system: HEK293 cells
  • Tag: C-terminal His tag
  • Function: participates in STAT3 activation via heterodimeric receptors
  • Applications: biochemical assays and cell-signaling studies
  • Form: recombinant protein

Catalog No. 50-002-90210 Supplier Medchemexpress LLC Supplier No. HYP764292UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant mouse interleukin-20 receptor subunit beta (IL-20RB) produced in HEK293 cells with a C-terminal His tag. IL-20RB forms functional heterodimers (for example with IL-20RA or IL-22RA1) and participates in STAT3 signaling; it serves as a receptor subunit for IL-19, IL-20, and IL-24. The protein is intended for biochemical characterization and cell-signaling assays.

  • Species: mouse
  • Expression system: HEK293 cells
  • Tag: C-terminal His tag
  • Function: participates in STAT3 activation via heterodimeric receptors
  • Applications: biochemical assays and cell-signaling studies
  • Form: recombinant protein

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.