Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Interleukin-21 (canine) | >95.0% | 10 UG
SDP

Catalog No. 5000292808
Encompass_Preferred

Recombinant canine interleukin-21 supplied as a lyophilized protein for research use. Expressed in E. coli and comprising amino acids H18-S146, it has an approximate molecular weight of 15 kDa and ≥95% purity. The product is formulated for stability and supplied in a 10 μg size suitable for small-scale assays.

  • Recombinant protein for immunology and cell signaling studies.
  • Produced in E. coli and provided as a lyophilized powder for stability.
  • Contains amino acids H18-S146 with an approximate molecular weight of 15 kDa.
  • Purity ≥95% as determined by reducing SDS-PAGE.
  • Formulated in PBS with trehalose, mannitol, and Tween-80 for preservation.
  • Available in a 10 μg size for small-scale experiments.
  • Store at -20°C and freeze aliquots at -20°C or -80°C for extended storage.

Catalog No. 50-002-92808 Supplier Medchemexpress LLC Supplier No. HYP7481910UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant canine interleukin-21 supplied as a lyophilized protein for research use. Expressed in E. coli and comprising amino acids H18-S146, it has an approximate molecular weight of 15 kDa and ≥95% purity. The product is formulated for stability and supplied in a 10 μg size suitable for small-scale assays.

  • Recombinant protein for immunology and cell signaling studies.
  • Produced in E. coli and provided as a lyophilized powder for stability.
  • Contains amino acids H18-S146 with an approximate molecular weight of 15 kDa.
  • Purity ≥95% as determined by reducing SDS-PAGE.
  • Formulated in PBS with trehalose, mannitol, and Tween-80 for preservation.
  • Available in a 10 μg size for small-scale experiments.
  • Store at -20°C and freeze aliquots at -20°C or -80°C for extended storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.