Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Interleukin-21 (canine), recombinant protein | >95.0% | 2 UG
SDP

Catalog No. 5000291540
Encompass_Preferred

Recombinant canine interleukin-21 (IL-21) supplied lyophilized as a tag-free protein expressed in E. coli for research use. It supports studies of humoral and cellular immunity and includes formulation, storage, and analytical purity data (sequence H18-S146; accession Q6L7I9).

  • Recombinant canine IL-21, tag-free
  • Expressed in E. coli
  • Sequence H18-S146; accession Q6L7I9
  • Purity >95.0% by reducing SDS-PAGE
  • Predicted mass ~15 kDa
  • Lyophilized with stabilizing excipients
  • Store at -20°C; reconstitute per datasheet (recommend ≥100 μg/mL)

Catalog No. 50-002-91540 Supplier Medchemexpress LLC Supplier No. HYP748192UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant canine interleukin-21 (IL-21) supplied lyophilized as a tag-free protein expressed in E. coli for research use. It supports studies of humoral and cellular immunity and includes formulation, storage, and analytical purity data (sequence H18-S146; accession Q6L7I9).

  • Recombinant canine IL-21, tag-free
  • Expressed in E. coli
  • Sequence H18-S146; accession Q6L7I9
  • Purity >95.0% by reducing SDS-PAGE
  • Predicted mass ~15 kDa
  • Lyophilized with stabilizing excipients
  • Store at -20°C; reconstitute per datasheet (recommend ≥100 μg/mL)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.