Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Interleukin-21 protein, rat | >95.0% | 500 UG
SDP

Catalog No. 5000293712
Encompass_Preferred

Recombinant rat interleukin-21 supplied lyophilized for research use, expressed in E. coli and provided as a 500 μg vial. Intended for functional assays and binding studies; handle and store frozen for extended stability.

  • Species: rat, recombinant protein
  • Source: expressed in E. coli
  • Formulation: lyophilized from PBS, pH 7.4
  • Amount: 500 μg per vial
  • Purity: >95.0% by reducing SDS-PAGE
  • Molecular weight: ~16 kDa (reducing SDS-PAGE)
  • Sequence range: P25-S146
  • Recommended use: functional ELISA; reported ED50 12.16 ng/mL
  • Storage: store at -20°C; freeze aliquots at -20°C or -80°C for long-term

Catalog No. 50-002-93712 Supplier Medchemexpress LLC Supplier No. HYP74818500UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant rat interleukin-21 supplied lyophilized for research use, expressed in E. coli and provided as a 500 μg vial. Intended for functional assays and binding studies; handle and store frozen for extended stability.

  • Species: rat, recombinant protein
  • Source: expressed in E. coli
  • Formulation: lyophilized from PBS, pH 7.4
  • Amount: 500 μg per vial
  • Purity: >95.0% by reducing SDS-PAGE
  • Molecular weight: ~16 kDa (reducing SDS-PAGE)
  • Sequence range: P25-S146
  • Recommended use: functional ELISA; reported ED50 12.16 ng/mL
  • Storage: store at -20°C; freeze aliquots at -20°C or -80°C for long-term

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.