Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Interleukin-21 (rat) | >95.0% | 10 UG
SDP

Catalog No. 5000293557
Encompass_Preferred

Recombinant rat interleukin-21 (partial, residues P25-S146) is supplied as a tag-free, E. coli-expressed lyophilized protein for research use in immunology and cell-signaling assays. The material is provided in a 10 μg unit and is characterized for purity, molecular mass, endotoxin level, and functional activity.

  • Expressed in E. coli; tag free.
  • Partial fragment P25-S146; sequence provided.
  • Purity >95.0% by reducing SDS-PAGE.
  • Observed molecular weight ~16 kDa (predicted 14.5 kDa).
  • Endotoxin <1 EU/μg (LAL method).
  • Biological activity: functional ELISA, ED50 = 12.16 ng/mL.
  • Formulation: lyophilized from PBS, pH 7.4.
  • Storage: store at -20°C; freeze aliquots for long-term stability.

Catalog No. 50-002-93557 Supplier Medchemexpress LLC Supplier No. HYP7481810UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant rat interleukin-21 (partial, residues P25-S146) is supplied as a tag-free, E. coli-expressed lyophilized protein for research use in immunology and cell-signaling assays. The material is provided in a 10 μg unit and is characterized for purity, molecular mass, endotoxin level, and functional activity.

  • Expressed in E. coli; tag free.
  • Partial fragment P25-S146; sequence provided.
  • Purity >95.0% by reducing SDS-PAGE.
  • Observed molecular weight ~16 kDa (predicted 14.5 kDa).
  • Endotoxin <1 EU/μg (LAL method).
  • Biological activity: functional ELISA, ED50 = 12.16 ng/mL.
  • Formulation: lyophilized from PBS, pH 7.4.
  • Storage: store at -20°C; freeze aliquots for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.