Learn More
Creative Biomart Interleukin-3 (IL-3) Protein
Catalog No. 502084285
Encompass_Preferred
Recombinant Human IL-3 is a 15.0 kDa globular protein containing 133 amino acid residues.
Catalog No. 50-208-4285 Supplier Creative Biomart Supplier No. THP0234
Recombinant Human IL-3 is a 15.0 kDa globular protein containing 133 amino acid residues.