Learn More
Sigma Aldrich Fine Chemicals Biosciences Interleukin 3 from mouse I50UG
Catalog No. 502914188
Encompass
Interleukin-3 is a hematopoietic growth factor which stimulates basophil development and triggers histamine IL-4 and IL-6 release. It improves monocyte and macrophage activation and phagocytosis enhances the secretory function of eosinophils. Interleukin-3 promotes endothelial cell proliferation and migration.
Catalog No. 50-291-4188 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. I414450UG
Interleukin-3 is a hematopoietic growth factor which stimulates basophil development and triggers histamine IL-4 and IL-6 release. It improves monocyte and macrophage activation and phagocytosis enhances the secretory function of eosinophils. Interleukin-3 promotes endothelial cell proliferation and migration.