Learn More
Cusabio Technology LLC Interleukin-36 alphaIL1F6
Catalog No. 5000049731
Encompass_Preferred
The Mouse Interleukin-36 alpha (IL1F6/Il36a) ELISA Kit is designed for the quantitative detection of Il36a in serum plasma and tissue homogenates using a sandwich ELISA technique This kit offers a broad detection range of 15 6 pg/mL to 1000 pg/mL with a high sensitivity of 3 9 pg/mL ensuring accurate measurement of low-abundance targets The assay requires a short incubation period of 1-5 hours and a sample volume of 50-100 L per test Detection is performed at 450 nm providing reliable and reproducible results for research applications Suitable for both routine laboratory use and specialized studies this kit maintains robust performance across validated sample types while adhering to standardized quantitative analysis protocols
Catalog No. 50-000-49731 Supplier Cusabio Technology LLC Supplier No. CSBEL011617MO
The Mouse Interleukin-36 alpha (IL1F6/Il36a) ELISA Kit is designed for the quantitative detection of Il36a in serum plasma and tissue homogenates using a sandwich ELISA technique This kit offers a broad detection range of 15 6 pg/mL to 1000 pg/mL with a high sensitivity of 3 9 pg/mL ensuring accurate measurement of low-abundance targets The assay requires a short incubation period of 1-5 hours and a sample volume of 50-100 L per test Detection is performed at 450 nm providing reliable and reproducible results for research applications Suitable for both routine laboratory use and specialized studies this kit maintains robust performance across validated sample types while adhering to standardized quantitative analysis protocols