Learn More
Medchemexpress LLC Interleukin-36 gamma | 50 UG
SDP
Catalog No. 5000292117
Encompass_Preferred
Recombinant human interleukin-36 gamma (IL-36γ, IL-1F9) is an animal-free, E. coli-expressed cytokine supplied as a lyophilized His-tagged protein. It activates NF-κB and MAPK signaling and induces chemokine expression; biological activity in A431 cells shows an ED50 <5 ng/mL. The product is formulated in PBS with trehalose, has low endotoxin, and is intended for cell-culture research use.
  • Lyophilized His-tagged recombinant human IL-36γ.
  • Expressed in Escherichia coli to provide animal-free material.
  • Approximate molecular weight ~19 kDa by SDS-PAGE.
  • Low endotoxin level (<0.1 EU per μg).
  • Demonstrated biological activity: ED50 <5 ng/mL (IL-8 induction in A431 cells).
  • Formulated in PBS, pH 8.0, with trehalose for stability.
  • Reconstitute to 100-200 μg/mL and aliquot to avoid freeze-thaw cycles.
  • For research use only; not for therapeutic or human use.
Catalog No. 50-002-92117 Supplier Medchemexpress LLC Supplier No. HYP75861AF50UG
Recombinant human interleukin-36 gamma (IL-36γ, IL-1F9) is an animal-free, E. coli-expressed cytokine supplied as a lyophilized His-tagged protein. It activates NF-κB and MAPK signaling and induces chemokine expression; biological activity in A431 cells shows an ED50 <5 ng/mL. The product is formulated in PBS with trehalose, has low endotoxin, and is intended for cell-culture research use.
  • Lyophilized His-tagged recombinant human IL-36γ.
  • Expressed in Escherichia coli to provide animal-free material.
  • Approximate molecular weight ~19 kDa by SDS-PAGE.
  • Low endotoxin level (<0.1 EU per μg).
  • Demonstrated biological activity: ED50 <5 ng/mL (IL-8 induction in A431 cells).
  • Formulated in PBS, pH 8.0, with trehalose for stability.
  • Reconstitute to 100-200 μg/mL and aliquot to avoid freeze-thaw cycles.
  • For research use only; not for therapeutic or human use.