Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Interleukin 4, human

Catalog No. AAJ67406YAM
Change view
Click to view available options
Quantity:
5 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ67406YAM 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ67406YAM Supplier Thermo Scientific Chemicals Supplier No. J67406YAM
Add to Cart

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Molecular Weight (g/mol) 15 kd
Quantity 5 μg
Packaging Plastic bottle
Form Lyophilized
How should I dilute Interleukin 4, human (Cat. No. J67406XXX) for use?

You should briefly centrifuge Interleukin 4, human (Cat. No. J67406XXX) prior to opening to bring the contents to the bottom. Reconstitute it in a siliconized tube to a final concentration of 0.1-1.0 mg/mL IL-4 using PBS containing 0.1% BSA. Reconstituted solutions are stable for up to one week at 2-8 degrees C. We recommend that you aliquot stock solutions and store the aliquots at -80 degrees C. Further dilutions should be made in appropriate buffered solutions containing BSA or serum.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.