Learn More
Cusabio Technology LLC interleukin 4 IL-4
Supplier:  Cusabio Technology LLC CSBE10126GU
Encompass_Preferred
This ELISA kit is designed for the quantitative detection of Guinea pig interleukin 4 (IL-4) in serum plasma and tissue homogenates using a sandwich assay methodology The kit demonstrates a detection range of 3 12 pg/mL to 200 pg/mL with a sensitivity of 0 78 pg/mL enabling precise measurement of low-abundance targets The procedure requires 50-100 L of sample per test and delivers results within 1-5 hours Absorbance is measured at 450 nm to determine IL-4 concentrations ensuring reliable performance across diverse biological matrices This kit detects IL-4 with high specificity and reproducibility making it suitable for research applications requiring accurate cytokine level analysis
Catalog No. 50-000-49476