Learn More
Sigma Aldrich Fine Chemicals Biosciences Interleukin-6 human IL-6 r
Catalog No. 502904122
Encompass
The IL6 (interleukin-6) gene is mapped to human chromosome 7p15.3. All the members of IL6 family have membrane glycoprotein gp130 as a common receptor. IL6 (interleukin-6) induces B-cell differentiation and is a primary regulator of inflammation.
Catalog No. 50-290-4122 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. H741610UG
The IL6 (interleukin-6) gene is mapped to human chromosome 7p15.3. All the members of IL6 family have membrane glycoprotein gp130 as a common receptor. IL6 (interleukin-6) induces B-cell differentiation and is a primary regulator of inflammation.