Learn More
Cayman Chemical InterleukIn-854-72humn 10mg
SDP
Catalog No. 502919358
Encompass_Preferred
A peptide; inhibits flow-based adhesion of primary human neutrophils to HUVECs
Catalog No. 50-291-9358 Supplier Cayman Chemical Supplier No. 3697810 MG
A peptide; inhibits flow-based adhesion of primary human neutrophils to HUVECs