Learn More
Cayman Chemical InterleukIn-854-72humn 5mg
SDP
Supplier:  Cayman Chemical 369785 MG
Encompass_Preferred
A peptide; inhibits flow-based adhesion of primary human neutrophils to HUVECs
Catalog No. 50-291-9359