Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Interleukin-9 receptor, rat (HEK293, His) | >95.0% | 10 UG
SDP

Catalog No. 5000290059
Encompass_Preferred

Recombinant rat interleukin-9 receptor extracellular domain expressed in HEK293 cells with a C-terminal 6xHis tag. The purified, glycosylated protein (sequence M1-A270) is provided at high purity for use in biochemical and biophysical assays, antibody validation, and receptor-ligand interaction studies. Supplied in small pack sizes for research use.

  • Expression system: HEK293
  • Tag: C-terminal 6xHis
  • Species: rat
  • Sequence: residues M1-A270 (270 aa)
  • Purity: >95.0% by reducing SDS-PAGE
  • Molecular weight: ~33-40 kDa (glycosylated)
  • Available pack sizes: includes 10 μg
  • Storage: store at -20°C; stable at 4°C for 1 week

Catalog No. 50-002-90059 Supplier Medchemexpress LLC Supplier No. HYP7643710UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant rat interleukin-9 receptor extracellular domain expressed in HEK293 cells with a C-terminal 6xHis tag. The purified, glycosylated protein (sequence M1-A270) is provided at high purity for use in biochemical and biophysical assays, antibody validation, and receptor-ligand interaction studies. Supplied in small pack sizes for research use.

  • Expression system: HEK293
  • Tag: C-terminal 6xHis
  • Species: rat
  • Sequence: residues M1-A270 (270 aa)
  • Purity: >95.0% by reducing SDS-PAGE
  • Molecular weight: ~33-40 kDa (glycosylated)
  • Available pack sizes: includes 10 μg
  • Storage: store at -20°C; stable at 4°C for 1 week

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.