Learn More
Apexbio Technology LLC β-Interleukin I (163-171), human 106021-96-9 10mg
SDP
Catalog No. 5000093300
Encompass_Preferred
beta-Interleukin I 163-171 human is a peptide fragment derived from human interleukin-1 (IL-1 ) It is utilized to investigate immune signaling pathways particularly those involved in inflammatory responses and T cell activation beta-Interleukin I 163-171 human exerts its biological activity by participating in the modulation of inflammatory signaling cascades relevant to immune system regulation Based on these pharmacological properties beta-Interleukin I 163-171 human holds research potential in the study of immune activation inflammation and associated signaling mechanisms in biomedical research
Catalog No. 50-000-93300 Supplier Apexbio Technology LLC Supplier No. A102610
beta-Interleukin I 163-171 human is a peptide fragment derived from human interleukin-1 (IL-1 ) It is utilized to investigate immune signaling pathways particularly those involved in inflammatory responses and T cell activation beta-Interleukin I 163-171 human exerts its biological activity by participating in the modulation of inflammatory signaling cascades relevant to immune system regulation Based on these pharmacological properties beta-Interleukin I 163-171 human holds research potential in the study of immune activation inflammation and associated signaling mechanisms in biomedical research