Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Interleukin (IL)-6 Receptor | 96.2% | 1216.30 | 5 MG
SDP

Supplier:  Medchemexpress LLC HYP03175MG

Encompass_Preferred

Interleukin (IL)-6 Receptor is a peptide, derived from interleukin-6 receptor. This product is intended for research use only and has not been fully validated for medical applications.

  • Appearance: Solid, white to off-white
  • Molecular formula: C51H85N13O21
  • Sequence: Thr-Ser-Leu-Pro-Val-Gln-Asp-Ser-Ser-Ser-Val-Pro
  • Soluble in water and PBS
  • Target: Interleukin related
  • Pathway: Immunology/Inflammation

Catalog No. 50-004-22143


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.