Learn More
Cayman Chemical InterleukIn1humn AChEF 500dtn
SDP
Catalog No. 502892440
Encompass_Preferred
A GPR52 agonist increases cAMP concentration in CHO cells expressing human GPR52 (EC50 75 nM) selective for GPR52 over 98 other receptors ion channels and enzymes at 10 UM increases c-Fos expression in the nucleus accumbens shell to a greater extent than in the dorsomedial striatum at 30 mg/kg increases the time spent with the novel object in the novel object recognition test in rats at 3 and 10 mg/kg
Catalog No. 50-289-2440 Supplier Cayman Chemical Supplier No. 483310500 DTN
A GPR52 agonist increases cAMP concentration in CHO cells expressing human GPR52 (EC50 75 nM) selective for GPR52 over 98 other receptors ion channels and enzymes at 10 UM increases c-Fos expression in the nucleus accumbens shell to a greater extent than in the dorsomedial striatum at 30 mg/kg increases the time spent with the novel object in the novel object recognition test in rats at 3 and 10 mg/kg