Learn More
Research Products International Corp Interlocking Cube Racks, Assorted Colors (Green, Yellow, Pink, Blue, Orange) 5 per Case
SDP
Catalog No. 50550319
Encompass
Now custom design your own tube rack system using our locking cube modules. Each cube interlocks on all sides for the exact configuration needed. Constructed of durable polypropylene. Available in assorted colors (green, yellow, pink, blue and orange).
Catalog No. 50-550-319 Supplier Research Products International Corp Supplier No. 248876
Now custom design your own tube rack system using our locking cube modules. Each cube interlocks on all sides for the exact configuration needed. Constructed of durable polypropylene. Available in assorted colors (green, yellow, pink, blue and orange).