Learn More
3M™ InterMask Face Masks
Catalog No. 19045571 Shop All 3M Products
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
19-045-571 1 Ea.
1 options
Catalog No. 19-045-571 Supplier 3M™ Supplier No. 7000126524
Add to Cart
Add to Cart
Attaches to 7800S/7800S-AM Series full facepiec respirators.
  • These safety products must be used in accordance with the user instructions, warnings and limitations accompanying each product.
  • 3M Catalog# 7990/7990-AM
TRUSTED_SUSTAINABILITY
Specifications
Product Type Clip-On Welding Face Shield Kit
For Use With (Equipment) 3m™ electronic lens assembly (7991/7991-am), full facepiece repirators (7800s), compatible with standard 4.5 x 5.25 in. lens
Includes Weld shield, cover plate, lens gasket
Model 7990/7990-AM
Quantity 1 Ea.