Learn More
Andwin Scientific INTERMED CRYMOLD 15/15/5 12/CS
SDP
Catalog No. NC9008138
Encompass_Preferred
The Cryomold molds produce uniformly shaped, flat-surface specimen block when used with O.C.T. (optimum cutting temperature) compound, which is sold separately, for frozen sectioning. This can fit into Cryobar freezing wells of the Tissue Tek II microtome/cryostat with the use of an adapter. This product measures 15 mm x 15 mm x 5 mm. It is sold in a case of 1200.
Catalog No. NC9008138 Supplier Andwin Scientific Supplier No. 4566CS
The Cryomold molds produce uniformly shaped, flat-surface specimen block when used with O.C.T. (optimum cutting temperature) compound, which is sold separately, for frozen sectioning. This can fit into Cryobar freezing wells of the Tissue Tek II microtome/cryostat with the use of an adapter. This product measures 15 mm x 15 mm x 5 mm. It is sold in a case of 1200.