Learn More
Bachem pTH (1-34) (human) Trifluoroacetate
SDP
Supplier:  Bachem 4095855.0500
Encompass_Preferred
pTH (1-34) (human) Trifluoroacetate, H-Ser-Val-Ser-Glu-Ile-Gln-Leu-Met-His-Asn-Leu-Gly-Lys-His-Leu-Asn-Ser-Met-Glu-Arg-Val-Glu-Trp-Leu-Arg-Lys-Lys-Leu-Gln-Asp-Val-His-Asn-Phe-OH, 52232-67-4, C181H291N55O51S2, Mr 4117.77, 0.5mg. Teriparatide, Parathyroid Hormone (1-34), human (Teriparatide). Intermittent administration of teriparatide stimulates bone formation by increasing the number of osteoblasts. Agonist of the parathyroid hormone receptor PTH1.
Catalog No. 50-194-6979