Learn More
Chemglass Life Sciences Internal Bushing, #50, PTFE, 1/4" NPT, #225 Viton O-Ring
SDP
Catalog No. 50873540
Encompass_Preferred
Internal Bushing, #50, PTFE, 1/4" NPT, #225 Viton O-Ring. New front seal PTFE bushing with female NPT for use with CG-360 internal threads. Supplied complete with Viton o-ring of the listed size. See CG-305 for Viton and CG-309 for Perfluoro replacement o-rings.
Catalog No. 50-873-540 Supplier Chemglass Life Sciences Supplier No. CG36452
Internal Bushing, #50, PTFE, 1/4" NPT, #225 Viton O-Ring. New front seal PTFE bushing with female NPT for use with CG-360 internal threads. Supplied complete with Viton o-ring of the listed size. See CG-305 for Viton and CG-309 for Perfluoro replacement o-rings.