Learn More
BIOHIPPO Internal Ctrl Lentivirus LV
SDP
Catalog No. 5000330836
Encompass
Internal Ctrl Lentivirus is a constitutively active control lentiviral vector used as a reference in transcription factor reporter assays The construct expresses a pathway independent reporter gene under a non responsive promoter so that signal reflects only infection efficiency and cell health It can be co transduced with pathway specific reporter lentivirus to normalize for differences in transduction rate cell number or viability The lentivirus is supplied as a ready to use VSV G pseudotyped preparation with a nominal titer of 5x10E6 transducing units TU This product is intended for research use only and is not for diagnostic or therapeutic procedures
Catalog No. 50-003-30836 Supplier BIOHIPPO Supplier No. BHV194000039
Internal Ctrl Lentivirus is a constitutively active control lentiviral vector used as a reference in transcription factor reporter assays The construct expresses a pathway independent reporter gene under a non responsive promoter so that signal reflects only infection efficiency and cell health It can be co transduced with pathway specific reporter lentivirus to normalize for differences in transduction rate cell number or viability The lentivirus is supplied as a ready to use VSV G pseudotyped preparation with a nominal titer of 5x10E6 transducing units TU This product is intended for research use only and is not for diagnostic or therapeutic procedures