Learn More
BIOHIPPO Internal Ctrl Lentivirus LV
SDP
Catalog No. 5000331653
Encompass
Internal Ctrl Lentivirus is a transcription factor reporter lentiviral vector designed to monitor transcription factor pathways in living cells This construct delivers a Renilla Luc reporter cassette together with a Blasticidin selection marker for generation of stable cell pools The vector is provided as a ready to use Lentivirus preparation pseudotyped with VSV-G for efficient gene delivery and stable genomic integration Each vial provides a nominal titer of 2x10E6 transducing units TU Typical applications include Monitor activity of ROR transcription factors The reporter could be useful for studying Th17 cell function The fluorescent reporter enables convenient readout using flow cytometry fluorescence microscopy etc The system is suitable for use with Human/mouse and can support assay development high content screening and mechanism of action studies This product is intended for research use only and is not for diagnostic or therapeutic procedures
Catalog No. 50-003-31653 Supplier BIOHIPPO Supplier No. BHV194000287
Internal Ctrl Lentivirus is a transcription factor reporter lentiviral vector designed to monitor transcription factor pathways in living cells This construct delivers a Renilla Luc reporter cassette together with a Blasticidin selection marker for generation of stable cell pools The vector is provided as a ready to use Lentivirus preparation pseudotyped with VSV-G for efficient gene delivery and stable genomic integration Each vial provides a nominal titer of 2x10E6 transducing units TU Typical applications include Monitor activity of ROR transcription factors The reporter could be useful for studying Th17 cell function The fluorescent reporter enables convenient readout using flow cytometry fluorescence microscopy etc The system is suitable for use with Human/mouse and can support assay development high content screening and mechanism of action studies This product is intended for research use only and is not for diagnostic or therapeutic procedures