Learn More
BIOHIPPO Internal Ctrl Lentivirus LV
SDP
Catalog No. 5000331699
Encompass
Internal Ctrl Lentivirus is a transcription factor reporter lentiviral vector designed to monitor the Retinoic acid signaling pathway Retinoid X Receptor signaling signaling pathway in living cells It delivers a Renilla Luc reporter cassette together with a Blasticidin selection marker for generation of stable cell pools The lentiviral vector pseudotyped with VSV-G enables efficient gene delivery and stable genomic integration Typical applications include pathway activity monitoring assay development high content screening and mechanism of action studies It is suitable for use with Human/mouse cells This product is intended for research use only and is not for diagnostic or therapeutic procedures
Catalog No. 50-003-31699 Supplier BIOHIPPO Supplier No. BHV194000437
Internal Ctrl Lentivirus is a transcription factor reporter lentiviral vector designed to monitor the Retinoic acid signaling pathway Retinoid X Receptor signaling signaling pathway in living cells It delivers a Renilla Luc reporter cassette together with a Blasticidin selection marker for generation of stable cell pools The lentiviral vector pseudotyped with VSV-G enables efficient gene delivery and stable genomic integration Typical applications include pathway activity monitoring assay development high content screening and mechanism of action studies It is suitable for use with Human/mouse cells This product is intended for research use only and is not for diagnostic or therapeutic procedures