Learn More
Surgical Design™ Internal Digital Caliper
Catalog No. 21124921 Shop All Surgical Design Products
Catalog No. 21-124-921 Supplier Surgical Design™ Supplier No. MT005
Add to Cart
Add to Cart
Internal digital caliper delivers accurate internal diameter measurements for laboratory, industrial, machining, and quality-control applications.
  • Designed for internal measurements including inner diameters, slots, grooves, and recesses
  • High-resolution digital readout provides fast, easy-to-read measurements
  • Hardened measuring jaws ensure consistent accuracy and long-term durability
  • Unit conversion (mm/in.) at the touch of a button for versatile workflow compatibility
  • Zero-setting and incremental measurement modes for comparative gauging
  • Smooth slide mechanism for precise control and repeatable results