Learn More
Trajan Scientific VICI Valco Internal Reducer, 1/16" to 1/32" Tubing OD, 0.25mm Bore, Without Frit
Catalog No. 501090218
Encompass
VICI Valco Internal Reducer, 1/16" to 1/32" Tubing OD, 0.25mm Bore, Without Frit. Made in the USA.
Catalog No. 50-109-0218 Supplier Trajan Scientific Supplier No. IZR1.5
VICI Valco Internal Reducer, 1/16" to 1/32" Tubing OD, 0.25mm Bore, Without Frit. Made in the USA.