Learn More
Hach Company Internal Relay Board
Supplier: Hach Company YAB076
Internal Relay Board for SC1000 Controller and 5500sc AnalyzersThis internal relay board provides 4 contact closure relays that can be used with the SC1000 Controller or 5500sc Analyzers