Learn More
AB Sciex LLC 10 kD marker for use with the SDS-MW Kit
Catalog No. NC1033092
Encompass
For research use only. Not for use in diagnostic procedures. 10 kD marker for use with the SDS-MW Kit (Part Number 390953) and the IgG Purity and Heterogeneity Assay Kit (Part Number A10663). Contains one tube of marker (400 ?L).
Catalog No. NC1033092 Supplier AB Sciex LLC Supplier No. A26487
For research use only. Not for use in diagnostic procedures. 10 kD marker for use with the SDS-MW Kit (Part Number 390953) and the IgG Purity and Heterogeneity Assay Kit (Part Number A10663). Contains one tube of marker (400 ?L).