Learn More
Perkin Elmer US LLC Internal Standard for Method 8260B
Supplier:  Perkin Elmer US LLC N9331041
Encompass_Preferred
Internal Standard for Method 8260B 4 certified analytes. 1. 2 mL @2000µg/ml - in P & T methanol2000 µg/ml - Chlorobenzene-d52000 µg/ml - 1,4-Dichlorobenzene-d42000 µg/ml - 1,4-Difluorobenzene2000 µg/ml - Pentafluorobenzene
Catalog No. 50-210-4679