Learn More
Glass Expansion Trident Internal Standard Kit
SDP
Catalog No. NC9281090
Encompass_Preferred
Trident Internal Standard Kit. In-line addition of internal standards & ionization buffers provides an efficient means of accurately & precisely dosing all of your samples without extra effort or the risk of error or contamination. Internal standardization (IS) is often used to compensate for physical & mass-space interferences in ICP spectrometry. It can also enhance short-term & long-term reproducibility. The Trident offers a more compact & efficient mixing chamber. Moreover, it is completely modular, so damaged or lost components sre easily replaced.Replacement parts include 60-808-1190, 60-808-1187, EZT-16-50T, UFT-EZT-16-50, EZT-16-50, EZT-16-25, NC1150995, KK711E.For the most recent design including ratchet connections use Glass Expansion’s Trident CT Internal Standard Kit (P/Ns NC2046960, 60-703-1179). For solutions containing HF use Trident IS Kit for HF P/N NC2046964, 60-808-1150.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. NC9281090 Supplier Glass Expansion Supplier No. 608081185
Trident Internal Standard Kit. In-line addition of internal standards & ionization buffers provides an efficient means of accurately & precisely dosing all of your samples without extra effort or the risk of error or contamination. Internal standardization (IS) is often used to compensate for physical & mass-space interferences in ICP spectrometry. It can also enhance short-term & long-term reproducibility. The Trident offers a more compact & efficient mixing chamber. Moreover, it is completely modular, so damaged or lost components sre easily replaced.Replacement parts include 60-808-1190, 60-808-1187, EZT-16-50T, UFT-EZT-16-50, EZT-16-50, EZT-16-25, NC1150995, KK711E.For the most recent design including ratchet connections use Glass Expansion’s Trident CT Internal Standard Kit (P/Ns NC2046960, 60-703-1179). For solutions containing HF use Trident IS Kit for HF P/N NC2046964, 60-808-1150.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001